2-Fluoro-5-pyridineboronic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20387-03
L20387-06
L20387-MD
CAAAAL20387-03EA
200.66
CAD
CAAAAL20387-03
CAAAAL20387-06
CAAAAL20387-MD
2-Fluoro-5-pyridineboronic acid 95%
2-Fluoro-5-pyridineboronic acid
Purity: 95%
CAS No.: 351019-18-6
Molecular Formula: C5H5BFNO2
Formula Weight: 140.91
Melting Point: 177-178° dec.
Beilstein Reference: 8974962
Formula:
C₅H₅BFNO₂ Melting Pt: 177…178 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD03411559 CAS Number: 351019-18-6 EINECS: 627-412-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...