(S)-(-)-3-Butyne-2-ol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L20345-03
L20345-06
CAAAAL20345-03EA
279.66
CAD
CAAAAL20345-03
CAAAAL20345-06
CAAAAL20345-MD
(S)-(-)-3-Butyne-2-ol 99%
(S)-(-)-3-Butyne-2-ol
Purity: 99%
CAS No.: 2914-69-4
Molecular Formula: C4H6O
Formula Weight: 70.09
Flash Point: 28°(82°F)
Refractive Index: 1.4260
Boiling Point: 108-109°
Beilstein Reference: 4652597
Optical Rotation: -47° (neat)
UN No.: 2929
MDL No.: MFCD00190166
Formula:
C₄H₆O Boiling Pt: 108…109 °C Density: 0.891 Flash Pt: 28 °C (82 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00190166 CAS Number: 2914-69-4 UN: 2929 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...