1,3-Dichloro-2-butene cis- and trans-mixture 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
L20334-06
L20334-14
CAAAAL20334-06EA
62.25
CAD
CAAAAL20334-06
CAAAAL20334-14
1,3-Dichloro-2-butene cis- and trans-mixture 98% stabilized
1,3-Dichloro-2-butene
Purity: 98%
CAS No.: 926-57-8
Molecular Formula: C4H6Cl2
Formula Weight: 125.00
Flash Point: 33°(91°F)
Refractive Index: 1.4730
Boiling Point: 125-129°
Beilstein Reference: 906754
UN No.: 1992
MDL No.: MFCD00000861
Formula:
C₄H₆Cl₂ MW: 125 g/mol Boiling Pt: 125…129 °C Melting Pt: –98 °C Density: 1.161 Flash Pt: 33 °C (91 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000861 CAS Number: 926-57-8 EINECS: 213-138-3 UN: 2920 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...