2,4-Dibromo-6-nitroaniline 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20333-06
L20333-14
CAAAAL20333-06EA
38.24
CAD
CAAAAL20333-06
CAAAAL20333-14
2,4-Dibromo-6-nitroaniline 99%
2,4-Dibromo-6-nitroaniline
Purity: 99%
CAS No.: 827-23-6
Molecular Formula: C6H4Br2N2O2
Formula Weight: 295.92
Melting Point: 129-131°
Beilstein Reference: 2805574
Formula:
C₆H₄Br₂N₂O₂ Melting Pt: 129…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012107 CAS Number: 827-23-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...