1,2,3-Trifluoro-5-nitrobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2,3-Trifluoro-5-nitrobenzene, 3,4,5-Trifluoronitrobenzene

L20302-06 L20302-14
CAAAAL20302-06EA 115.18 CAD
CAAAAL20302-06 CAAAAL20302-14
1,2,3-Trifluoro-5-nitrobenzene 99%
1,2,3-Trifluoro-5-nitrobenzene

Purity: 99% CAS No.: 66684-58-0 Molecular Formula: C6H2F3NO2 Formula Weight: 177.08 Flash Point: 75°(167°F) Refractive Index: 1.4860 Beilstein Reference: 2455018 UN No.: 2810 MDL No.: MFCD00456803

Formula: C₆H₂F₃NO₂
MW: 177.08 g/mol
Density: 1.52
Flash Pt: 75 °C (167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00456803
CAS Number: 66684-58-0
EINECS: 266-447-0
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR