Cerium(IV) sulfate, anhydrous 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ceric sulfate, Cerium disulfate, Cerium disulphate, Ceric sulphate, Cerium(IV) sulfate, Cerium(IV) sulphate

L20254-14 L20254-22 L20254-36
CAAAAL20254-14EA 102.23 CAD
CAAAAL20254-14 CAAAAL20254-22 CAAAAL20254-36
Cerium(IV) sulfate, anhydrous 97%
Cerium (IV) sulphate

Purity: 97% CAS No.: 13590-82-4 Molecular Formula: Ce(SO4)2 Formula Weight: 332.24 Merck Reference: 14,1990 UN No.: 3085 MDL No.: MFCD00148852

Formula: Ce(SO₄)₂
MW: 332.24 g/mol
Melting Pt: 195
Density: 3.91
Storage Temperature: Ambient
MDL Number: MFCD00148852
CAS Number: 13590-82-4
EINECS: 237-029-5
UN: 3260
ADR: 8 III
Merck Index: 14,1990

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR