Triethylsilylacetylene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L20217-03
L20217-06
CAAAAL20217-03EA
50.24
CAD
CAAAAL20217-03
CAAAAL20217-06
Triethylsilylacetylene 97%
Triethylsilylacetylene
Purity: 97%
CAS No.: 1777-03-3
Molecular Formula: HC≡CSi(CH2CH3)3
Formula Weight: 140.31
Flash Point: 17°(62°F)
Refractive Index: 1.4315
Boiling Point: 136-140°
Beilstein Reference: 1743814
UN No.: 1993
MDL No.: MFCD00075373
Formula:
C₈H₁₆Si Boiling Pt: 136…140 °C Density: 0.783 Flash Pt: 17 °C (62 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00075373 CAS Number: 1777-03-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...