Tributylvinyltin 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tri-n-butylvinyltin, Tributylvinylstannane

L20215-03 L20215-06
CAAAAL20215-03EA 71.57 CAD
CAAAAL20215-03 CAAAAL20215-06
Tributylvinyltin 96%
Tributylvinyltin

Purity: 96% CAS No.: 7486-35-3 Molecular Formula: C14H30Sn Formula Weight: 317.11 Flash Point: 110°(230°F) Refractive Index: 1.4780 Boiling Point: 104-106°/3.5mm Beilstein Reference: 3537662 UN No.: 2788 MDL No.: MFCD00009421

Formula: C₁₄H₃₀Sn
MW: 317.1 g/mol
Boiling Pt: 104…106 °C/3.5 mm
Density: 1.085
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009421
CAS Number: 7486-35-3
EINECS: 231-291-4
UN: 2788
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR