4-Quinolinecarbaldehyde 97+%

Supplier: Thermo Scientific Chemicals
Warning

L20203-06 L20203-14
CAAAAL20203-06EA 140.78 CAD
CAAAAL20203-06 CAAAAL20203-14
4-Quinolinecarbaldehyde 97+%
4-Quinolinecarbaldehyde

Purity: 97+% CAS No.: 4363-93-3 Molecular Formula: C10H7NO Formula Weight: 157.17 Flash Point: 110°(230°F) Melting Point: 48-52° Boiling Point: 131-133°/5mm Beilstein Reference: 113072 MDL No.: MFCD00006781

Formula: C₁₀H₇NO
Boiling Pt: 131…133 °C/5 mm
Melting Pt: 48…52 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006781
CAS Number: 4363-93-3
EINECS: 224-453-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR