2-Quinolinecarbaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Quinoline-2-carboxaldehyde, Quinoline-2-carbaldehyde, 2-Quinolinecarboxaldehyde

L20202-06 L20202-14
CAAAAL20202-06EA 121.66 CAD
CAAAAL20202-06 CAAAAL20202-14
2-Quinolinecarbaldehyde 97%
2-Quinolinecarbaldehyde

Purity: 97% CAS No.: 5470-96-2 Molecular Formula: C10H7NO Formula Weight: 157.17 Melting Point: 66-70° Beilstein Reference: 113061

Formula: C₁₀H₇NO
Melting Pt: 66…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00075032
CAS Number: 5470-96-2
EINECS: 226-804-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR