4-Iodoimidazole 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20184-03
L20184-06
CAAAAL20184-03EA
80.9
CAD
CAAAAL20184-03
CAAAAL20184-06
CAAAAL20184-MD
4-Iodoimidazole 94%
4-Iodoimidazole
Purity: 94%
CAS No.: 71759-89-2
Molecular Formula: C3H3IN2
Formula Weight: 193.96
Melting Point: 135-138°
Beilstein Reference: 1616
Formula:
C₃H₃IN₂ Melting Pt: 135…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD01632213 CAS Number: 71759-89-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...