6-Chloro-m-anisidine 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Chloro-3-anisidine, 2-Chloro-5-methoxyaniline, 3-Amino-4-chloroanisole

L20162-06 L20162-14 L20162-22
CAAAAL20162-06EA 100.96 CAD
CAAAAL20162-06 CAAAAL20162-14 CAAAAL20162-22
6-Chloro-m-anisidine 98+%
6-Chloro-m-anisidine

Purity: 98+% CAS No.: 2401-24-3 Molecular Formula: C7H8ClNO Formula Weight: 157.60 Refractive Index: 1.5865 Odor: Characteristic Boiling Point: 135-137°/12mm Beilstein Reference: 2082193 UN No.: 2233 MDL No.: MFCD00047830

Formula: C₇H₈ClNO
MW: 157.6 g/mol
Boiling Pt: 135…137 °C/12 mm
Melting Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00047830
CAS Number: 2401-24-3
EINECS: 219-277-6
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR