2-Chloro-5-iodobenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20161-06
L20161-14
L20161-22
CAAAAL20161-06EA
66.36
CAD
CAAAAL20161-06
CAAAAL20161-14
CAAAAL20161-22
2-Chloro-5-iodobenzoic acid 97%
2-Chloro-5-iodobenzoic acid
Purity: 97%
CAS No.: 19094-56-5
Molecular Formula: C7H4ClIO2
Formula Weight: 282.46
Melting Point: 156-161°
Beilstein Reference: 2254110
Formula:
C₇H₄ClIO₂ MW: 282.46 g/mol Melting Pt: 156…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD00079731 CAS Number: 19094-56-5 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...