4-Chloro-2-fluorobenzonitrile 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20160-03
L20160-06
L20160-14
CAAAAL20160-03EA
32.55
CAD
CAAAAL20160-03
CAAAAL20160-06
CAAAAL20160-14
4-Chloro-2-fluorobenzonitrile 98+%
4-Chloro-2-fluorobenzonitrile
Purity: 98+%
CAS No.: 57381-51-8
Molecular Formula: C7H3ClFN
Formula Weight: 155.56
Melting Point: 58-61°
Beilstein Reference: 4231142
UN No.: 3439
MDL No.: MFCD00143284
Formula:
C₇H₃ClFN Boiling Pt: 237 °C (1013 hPa) Melting Pt: 58…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143284 CAS Number: 57381-51-8 EINECS: 260-712-4 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...