6-Bromo-o-toluic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20149-03
L20149-06
CAAAAL20149-03EA
200.66
CAD
CAAAAL20149-03
CAAAAL20149-06
CAAAAL20149-MD
6-Bromo-o-toluic acid 98%
6-Bromo-o-toluic acid
Purity: 98%
CAS No.: 90259-31-7
Molecular Formula: C8H7BrO2
Formula Weight: 215.05
Melting Point: 107-111°
Beilstein Reference: 2614094
Formula:
C₈H₇BrO₂ Melting Pt: 107…111 °C Storage Temperature: Ambient |
MDL Number:
MFCD01310788 CAS Number: 90259-31-7 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...