2-Bromo-4-fluoro-1-iodobenzene 98+%

Supplier: Thermo Scientific Chemicals
Warning

L20144-03 L20144-06
CAAAAL20144-03EA 27.81 CAD
CAAAAL20144-03 CAAAAL20144-06
2-Bromo-4-fluoro-1-iodobenzene 98+%
2-Bromo-4-fluoro-1-iodobenzene

Purity: 98+% CAS No.: 202865-73-4 Molecular Formula: C6H3BrFI Formula Weight: 300.90 Flash Point: 110°(230°F) Refractive Index: 1.6290 Boiling Point: 224-225° Beilstein Reference: 6288420 MDL No.: MFCD00142868

Formula: C₆H₃BrFI
Boiling Pt: 224…225 °C
Density: 1.33
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00142868
CAS Number: 202865-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR