4-Bromo-2-fluorobenzoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L20142-03
L20142-06
L20142-14
CAAAAL20142-03EA
59.41
CAD
CAAAAL20142-03
CAAAAL20142-06
CAAAAL20142-14
4-Bromo-2-fluorobenzoyl chloride 98%
4-Bromo-2-fluorobenzoyl chloride
Purity: 98%
CAS No.: 151982-51-3
Molecular Formula: C7H3BrClFO
Formula Weight: 237.45
Refractive Index: 1.5877
Melting Point: 24-25°
Boiling Point: 86-88°/15mm
Beilstein Reference: 6323925
UN No.: 3261
MDL No.: MFCD03411584
Formula:
C₇H₃BrClFO Boiling Pt: 86…88 °C/15 mm Melting Pt: 24…25 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411584 CAS Number: 151982-51-3 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...