4-Amino-2,3,5,6-tetrafluorobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amino-2,3,5,6-tetrafluorobenzoic Acid

L20128-03 L20128-06
CAAAAL20128-03EA 74.1 CAD
CAAAAL20128-03 CAAAAL20128-06 CAAAAL20128-14
4-Amino-2,3,5,6-tetrafluorobenzoic acid 98%
4-Amino-2,3,5,6-tetrafluorobenzoic acid

Purity: 98% CAS No.: 944-43-4 Molecular Formula: C7H3F4NO2 Formula Weight: 209.11 Melting Point: 182-184° Beilstein Reference: 2121117

Formula: C₇H₃F₄NO₂
MW: 209.1 g/mol
Melting Pt: 182…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00007647
CAS Number: 944-43-4
EINECS: 213-409-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR