3-Ethynylaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Amino-3-ethynylbenzene, (3-Aminophenyl)acetylene, (3-Aminophenyl)ethyne, 1-Ethynyl-3-aminobenzene

L20124-06 L20124-14
CAAAAL20124-06EA 125.14 CAD
CAAAAL20124-06 CAAAAL20124-14
3-Ethynylaniline 98%
3-Ethynylaniline

Purity: 98% CAS No.: 54060-30-9 Molecular Formula: C8H7N Formula Weight: 117.15 Flash Point: 100°(212°F) Refractive Index: 1.6170 Melting Point: 27° Boiling Point: 70°/1mm Beilstein Reference: 2935417 MDL No.: MFCD00014779

Formula: C₈H₇N
MW: 117.15 g/mol
Boiling Pt: 50
Melting Pt: 27
Density: 1.040
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014779
CAS Number: 54060-30-9
EINECS: 258-944-6
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR