4-Chloro-3-(trifluoromethyl)phenylboronic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

L20105-03 L20105-06
CAAAAL20105-03EA 129.09 CAD
CAAAAL20105-03 CAAAAL20105-06
4-Chloro-3-(trifluoromethyl)phenylboronic acid 96%
4-Chloro-3-(trifluoromethyl)phenylboronic acid

Purity: 96% CAS No.: 176976-42-4 Molecular Formula: C7H5BClF3O2 Formula Weight: 224.38 MDL No.: MFCD03094999

Formula: C₇H₅BClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD03094999
CAS Number: 176976-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR