2-Chloro-5-(trifluoromethyl)phenylboronic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chloro-5-(trifluoromethyl)phenylboronic Acid, Dihydroxy(2-chloro-5-(trifluoromethyl)phenyl)borane, (2-Chloro-5-(trifluoromethyl)phenyl)metaboric acid, 2-Chloro-5-(trifluoromethyl)benzeneboronic acid, 2-Chloro-5-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride)
L20104-03
L20104-06
CAAAAL20104-03EA
68.57
CAD
CAAAAL20104-03
CAAAAL20104-06
2-Chloro-5-(trifluoromethyl)phenylboronic acid 96%
2-Chloro-5-(trifluoromethyl)phenylboronic acid
Purity: 96%
CAS No.: 182344-18-9
Molecular Formula: C7H5BClF3O2
Formula Weight: 224.38
MDL No.: MFCD00797335
Formula:
C₇H₅BClF₃O₂ MW: 224.37 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00797335 CAS Number: 182344-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...