2-Chloro-4-(trifluoromethyl)phenylboronic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Chloro-4-(trifluoromethyl)phenyl)metaboric acid, Dihydroxy(2-chloro-4-(trifluoromethyl)phenyl)borane, 2-Chloro-4-(trifluoromethyl)benzeneboronic acid

L20103-03 L20103-06
CAAAAL20103-03EA 75.21 CAD
CAAAAL20103-03 CAAAAL20103-06
2-Chloro-4-(trifluoromethyl)phenylboronic acid 96%
2-Chloro-4-(trifluoromethyl)phenylboronic acid

Purity: 96% CAS No.: 254993-59-4 Molecular Formula: C7H5BClF3O2 Formula Weight: 224.38 Melting Point: 110-112° Beilstein Reference: 9263388

Formula: C₇H₅BClF₃O₂
MW: 224.37
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD02684337
CAS Number: 254993-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR