2-Bromo-5-pyridineboronic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20085-03
L20085-06
L20085-MD
CAAAAL20085-03EA
265.44
CAD
CAAAAL20085-03
CAAAAL20085-06
CAAAAL20085-MD
2-Bromo-5-pyridineboronic acid 95%
2-Bromo-5-pyridineboronic acid
Purity: 95%
CAS No.: 223463-14-7
Molecular Formula: C5H5BBrNO2
Formula Weight: 201.82
Melting Point: ca 165° dec.
Beilstein Reference: 9119918
Formula:
C₅H₅BBrNO₂ Melting Pt: ∼ 165 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD03411558 CAS Number: 223463-14-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...