4-Chloronicotinic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chloro-3-pyridinecarboxylic acid, 4-Chloropyridine-3-carboxylic acid

L20029-03 L20029-06
CAAAAL20029-03EA 124.5 CAD
CAAAAL20029-03 CAAAAL20029-06
4-Chloronicotinic acid 97%
4-Chloronicotinic acid

Purity: 97% CAS No.: 10177-29-4 Molecular Formula: C6H4ClNO2 Formula Weight: 157.56 Melting Point: ca 165° dec. Beilstein Reference: 9255849

Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Melting Pt: ∼ 165 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00128860
CAS Number: 10177-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR