3-Chloro-2-hydroxypyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Chloro-2-pyridinol
L20027-03
L20027-06
L20027-14
CAAAAL20027-03EA
44.24
CAD
CAAAAL20027-03
CAAAAL20027-06
CAAAAL20027-14
3-Chloro-2-hydroxypyridine 97%
3-Chloro-2-hydroxypyridine
Purity: 97%
CAS No.: 13466-35-8
Molecular Formula: C5H4ClNO
Formula Weight: 129.55
Melting Point: 180-182°
Beilstein Reference: 109845
Formula:
C₅H₄ClNO MW: 129.55 g/mol Melting Pt: 180…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD01646109 CAS Number: 13466-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...