2-Bromo-3-nitropyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20019-03
L20019-06
CAAAAL20019-03EA
46.45
CAD
CAAAAL20019-03
CAAAAL20019-06
2-Bromo-3-nitropyridine 98%
2-Bromo-3-nitropyridine
Purity: 98%
CAS No.: 19755-53-4
Molecular Formula: C5H3BrN2O2
Formula Weight: 203.00
Melting Point: 122-125°
Beilstein Reference: 124504
Formula:
C₅H₃BrN₂O₂ Melting Pt: 122…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00955613 CAS Number: 19755-53-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...