2-Bromo-6-methylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20018-06
L20018-14
CAAAAL20018-06EA
117.24
CAD
CAAAAL20018-06
CAAAAL20018-14
2-Bromo-6-methylpyridine 98%
2-Bromo-6-methylpyridine
Purity: 98%
CAS No.: 5315-25-3
Molecular Formula: C6H6BrN
Formula Weight: 172.03
Flash Point: 97°(206°F)
Refractive Index: 1.5625
Boiling Point: 102-103°/20mm
Beilstein Reference: 107322
MDL No.: MFCD00040743
Formula:
C₆H₆BrN Boiling Pt: 102…103 °C/20 mm Density: 1.512 Flash Pt: 97 °C(206 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040743 CAS Number: 5315-25-3 EINECS: 226-173-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...