5-Bromo-2-methoxypyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20017-06
L20017-14
CAAAAL20017-06EA
205.4
CAD
CAAAAL20017-06
CAAAAL20017-14
5-Bromo-2-methoxypyridine 98%
5-Bromo-2-methoxypyridine
Purity: 98%
CAS No.: 13472-85-0
Molecular Formula: C6H6BrNO
Formula Weight: 188.02
Flash Point: 96°(204°F)
Refractive Index: 1.5550
Boiling Point: 80°/12mm
Beilstein Reference: 115150
MDL No.: MFCD01318952
Formula:
C₆H₆BrNO Boiling Pt: 80 °C/12 mm Density: 1.453 Flash Pt: 96 °C(204 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01318952 CAS Number: 13472-85-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...