2-Bromo-3-chloropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

L20012-03 L20012-06
CAAAAL20012-03EA 38.71 CAD
CAAAAL20012-03 CAAAAL20012-06
2-Bromo-3-chloropyridine 97%
2-Bromo-3-chloropyridine

Purity: 97% CAS No.: 96424-68-9 Molecular Formula: C5H3BrClN Formula Weight: 192.44 Melting Point: 59-61° Beilstein Reference: 1561638

Formula: C₅H₃BrClN
MW: 192.44 g/mol
Melting Pt: 59…61 °C
Storage Temperature: Ambient
MDL Number: MFCD04039344
CAS Number: 96424-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR