2-Amino-5-fluoropyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20010-03
L20010-06
CAAAAL20010-03EA
165.9
CAD
CAAAAL20010-03
CAAAAL20010-06
CAAAAL20010-MD
2-Amino-5-fluoropyridine 97%
2-Amino-5-fluoropyridine
Purity: 97%
CAS No.: 21717-96-4
Molecular Formula: C5H5FN2
Formula Weight: 112.11
Odor: Characteristic
Melting Point: 93-97°
Beilstein Reference: 385938
UN No.: 2811
MDL No.: MFCD01861120
Formula:
C₅H₅FN₂ Melting Pt: 93…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD01861120 CAS Number: 21717-96-4 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...