5-Amino-2-bromopyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Bromopyridin-3-amine, 2-Bromopyridin-5-amine, 3-Amino-6-bromopyridine, 2-Bromo-5-pyridylamine

L20008-06 L20008-14
CAAAAL20008-03EA 120.63 CAD
CAAAAL20008-03 CAAAAL20008-06 CAAAAL20008-14
5-Amino-2-bromopyridine 97%
5-Amino-2-bromopyridine

Purity: 97% CAS No.: 13534-97-9 Molecular Formula: C5H5BrN2 Formula Weight: 173.01 Melting Point: 75-79° Beilstein Reference: 109102

Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD01646116
CAS Number: 13534-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR