4-Amino-2-bromopyridine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L20007-03
L20007-06
CAAAAL20007-03EA
54.83
CAD
CAAAAL20007-03
CAAAAL20007-06
4-Amino-2-bromopyridine 97%
4-Amino-2-bromopyridine
Purity: 97%
CAS No.: 7598-35-8
Molecular Formula: C5H5BrN2
Formula Weight: 173.01
Melting Point: 96-99°
Beilstein Reference: 108928
Formula:
C₅H₅BrN₂ Melting Pt: 96…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD01646061 CAS Number: 7598-35-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...