2-(2-Fluorophenyl)ethanol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Fluorophenethyl alcohol
L19998-06
L19998-14
CAAAAL19998-06EA
113.44
CAD
CAAAAL19998-06
CAAAAL19998-14
2-(2-Fluorophenyl)ethanol 99%
2-(2-Fluorophenyl)ethanol
Purity: 99%
CAS No.: 50919-06-7
Molecular Formula: C8H9FO
Formula Weight: 140.16
Flash Point: 98°(208°F)
Refractive Index: 1.5100
Boiling Point: 72°/1mm
MDL No.: MFCD00002887
Formula:
C₈H₉FO MW: 140.16 g/mol Boiling Pt: 72 °C/1 mm Density: 1.045 Flash Pt: 98 °C (208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002887 CAS Number: 50919-06-7 UN: 2810 ADR: 6.1,III |
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...