1-(4-Nitrophenyl)glycerol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: PNPG, 1-(4-Nitrophenyl)-1,2,3-propanetriol

L19968-03 L19968-06
CAAAAL19968-03EA 282.82 CAD
CAAAAL19968-03 CAAAAL19968-06 CAAAAL19968-MD
1-(4-Nitrophenyl)glycerol 99%
1-(4-Nitrophenyl)glycerol

Purity: 99% CAS No.: 2207-68-3 Molecular Formula: C9H11NO5 Formula Weight: 213.19 Odor: Odorless Melting Point: 95-97° Beilstein Reference: 7376091 MDL No.: MFCD00017037

Formula: C₉H₁₁NO₅
MW: 213.19 g/mol
Melting Pt: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00017037
CAS Number: 2207-68-3
EINECS: 218-624-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR