1-Propylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

L19965-03 L19965-06 L19965-14
CAAAAL19965-03EA 79 CAD
CAAAAL19965-03 CAAAAL19965-06 CAAAAL19965-14
1-Propylboronic acid 98%
1-Propylboronic acid

Purity: 98% CAS No.: 17745-45-8 Molecular Formula: CH3(CH2)2B(OH)2 Formula Weight: 87.91 Storage Temperature: 2°C to 8°C Melting Point: 105-107° Beilstein Reference: 1731884 MDL No.: MFCD01074564

Formula: C₃H₉BO₂
Melting Pt: 105…107 °C
Storage Temperature: Ambient
MDL Number: MFCD01074564
CAS Number: 17745-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR