1-Octylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

L19964-03 L19964-06 L19964-14
CAAAAL19964-03EA 44.08 CAD
CAAAAL19964-03 CAAAAL19964-06 CAAAAL19964-14
1-Octylboronic acid 97%
1-Octylboronic acid

Purity: 97% CAS No.: 28741-08-4 Molecular Formula: CH3(CH2)7B(OH)2 Formula Weight: 158.05 Melting Point: 81-85° Beilstein Reference: 1739759

Formula: C₈H₁₉BO₂
Melting Pt: 81…85 °C
Storage Temperature: Ambient
MDL Number: MFCD01074560
CAS Number: 28741-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR