2-Nitrophenylboronic acid pinacol ester 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Nitrophenylboronic acid Pinacol ester, 2-Nitrobenzeneboronic acid pinacol ester, 2-Nitrophenylboronic acid, pinacol ester, 4,4,5,5-Tetramethyl-2-(2-nitrophenyl)-1,3,2-dioxaborolane, 1-Nitro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
L19963-03
L19963-06
CAAAAL19963-03EA
106.02
CAD
CAAAAL19963-03
CAAAAL19963-06
2-Nitrophenylboronic acid pinacol ester 98+%
2-Nitrophenylboronic acid pinacol ester
Purity: 98+%
CAS No.: 190788-59-1
Molecular Formula: C12H16BNO4
Formula Weight: 249.08
Melting Point: 42-44°
Beilstein Reference: 7708824
Formula:
C₁₂H₁₆BNO₄ MW: 249.07 g/mol Melting Pt: 42…44 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179447 CAS Number: 190788-59-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...