Ethylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19959-03
L19959-06
L19959-14
CAAAAL19959-03EA
76.63
CAD
CAAAAL19959-03
CAAAAL19959-06
CAAAAL19959-14
Ethylboronic acid 98%
Ethylboronic acid
Purity: 98%
CAS No.: 4433-63-0
Molecular Formula: C2H7BO2
Formula Weight: 73.89
Storage Temperature: 2°C to 8°C
Odor: Odorless
Beilstein Reference: 1731546
MDL No.: MFCD01074536
Formula:
C₂H₇BO₂ Melting Pt: 84…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD01074536 CAS Number: 4433-63-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...