1-Decylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19957-03
L19957-06
L19957-14
CAAAAL19957-03EA
70.47
CAD
CAAAAL19957-03
CAAAAL19957-06
CAAAAL19957-14
1-Decylboronic acid 98%
1-Decylboronic acid
Purity: 98%
CAS No.: 24464-63-9
Molecular Formula: CH3(CH2)9B(OH)2
Formula Weight: 186.10
Melting Point: 86-89°
Beilstein Reference: 1743934
Formula:
C₁₀H₂₃BO₂ Melting Pt: 86…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD01074638 CAS Number: 24464-63-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...