4-(Bromomethyl)phenylboronic acid tech. 85%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(4-(Bromomethyl)phenyl)metaboric acid, (4-(Bromomethyl)phenyl)boronic acid, Dihydroxy(4-(bromomethyl)phenyl)borane, 4-Bromomethylphenylboronic acid, 4-(Bromomethyl)benzeneboronic acid
L19953-03
L19953-06
CAAAAL19953-03EA
140.94
CAD
CAAAAL19953-03
CAAAAL19953-06
4-(Bromomethyl)phenylboronic acid tech. 85%, Technical Grade
4-(Bromomethyl)phenylboronic acid
Purity: 85%
CAS No.: 68162-47-0
Molecular Formula: C7H8BBrO2
Formula Weight: 214.85
Melting Point: ca 145-150°
Beilstein Reference: 3030979
UN No.: 3261
MDL No.: MFCD01318113
Formula:
C₇H₈BBrO₂ MW: 214.85 g/mol Melting Pt: ∼ 145…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318113 CAS Number: 68162-47-0 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...