2-Aminophenylboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 2-Aminobenzeneboronic acid pinacol ester
L19951-03
L19951-06
CAAAAL19951-03EA
83.58
CAD
CAAAAL19951-03
CAAAAL19951-06
2-Aminophenylboronic acid pinacol ester 97%
2-Aminophenylboronic acid pinacol ester
Purity: 97%
CAS No.: 191171-55-8
Molecular Formula: C12H18BNO2
Formula Weight: 219.09
Melting Point: 65-70°
Beilstein Reference: 8548809
Formula:
C₁₂H₁₈BNO₂ MW: 219.09 g/mol Melting Pt: 65…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179448 CAS Number: 191171-55-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...