3-(Trifluoromethoxy)thiophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

L19912-03 L19912-06
CAAAAL19912-03EA 175.38 CAD
CAAAAL19912-03 CAAAAL19912-06
3-(Trifluoromethoxy)thiophenol 98%
3-(Trifluoromethoxy)thiophenol

Purity: 98% CAS No.: 220239-66-7 Molecular Formula: C7H5F3OS Formula Weight: 194.18 Refractive Index: 1.4770

Formula: C₇H₅F₃OS
Storage Temperature: Ambient
MDL Number: MFCD01320799
CAS Number: 220239-66-7
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR