2-Methoxy-4-nitrobenzenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Methoxy-4-nitrobenzene-1-sulphonyl chloride, 2-Methoxy-4-nitrobenzene-1-sulfonyl chloride

L19902-03 L19902-06
CAAAAL19902-03EA 34.6 CAD
CAAAAL19902-03 CAAAAL19902-06
2-Methoxy-4-nitrobenzenesulfonyl chloride 97%
2-Methoxy-4-nitrobenzenesulphonyl chloride

Purity: 97% CAS No.: 21320-91-2 Molecular Formula: C7H6ClNO5S Formula Weight: 251.65 Melting Point: 96-99° Beilstein Reference: 2136023 UN No.: 3261 MDL No.: MFCD03094697

Formula: C₇H₆ClNO₅S
Melting Pt: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD03094697
CAS Number: 21320-91-2
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR