2,5-Dibromobenzenesulfonyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,5-Dibromobenzene-1-sulphonyl chloride, 2,5-Dibromobenzene-1-sulfonyl chloride

L19888-03 L19888-06
CAAAAL19888-03EA 101.59 CAD
CAAAAL19888-03 CAAAAL19888-06
2,5-Dibromobenzenesulfonyl chloride 99%
2,5-Dibromobenzenesulphonyl chloride

Purity: 99% CAS No.: 23886-64-8 Molecular Formula: C6H3Br2ClO2S Formula Weight: 334.41 Melting Point: 69-72° Beilstein Reference: 2694054 UN No.: 3261 MDL No.: MFCD00060685

Formula: C₆H₃Br₂ClO₂S
MW: 334.42 g/mol
Melting Pt: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00060685
CAS Number: 23886-64-8
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR