2,4-Dibromobenzenesulfonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,4-Dibromobenzene-1-sulfonyl chloride, 2,4-Dibromobenzene-1-sulphonyl chloride
L19887-03
L19887-06
CAAAAL19887-03EA
62.25
CAD
CAAAAL19887-03
CAAAAL19887-06
2,4-Dibromobenzenesulfonyl chloride 98%
2,4-Dibromobenzenesulphonyl chloride
Purity: 98%
CAS No.: 72256-95-2
Molecular Formula: C6H3Br2ClO2S
Formula Weight: 334.41
Melting Point: 82-86°
Boiling Point: 128°/0.2mm
UN No.: 3261
MDL No.: MFCD03093759
Formula:
C₆H₃Br₂ClO₂S MW: 334.42 g/mol Boiling Pt: 128 °C/0.2 mm Melting Pt: 82…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD03093759 CAS Number: 72256-95-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...