4-Bromo-2,6-dichlorobenzenesulfonyl chloride 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Bromo-2,6-dichlorobenzene-1-sulfonyl chloride, 4-Bromo-2,6-dichlorobenzene-1-sulphonyl chloride

L19879-03 L19879-06
CAAAAL19879-03EA 51.82 CAD
CAAAAL19879-03 CAAAAL19879-06
4-Bromo-2,6-dichlorobenzenesulfonyl chloride 95%
4-Bromo-2,6-dichlorobenzenesulphonyl chloride

Purity: 95% CAS No.: 351003-54-8 Molecular Formula: C6H2BrCl3O2S Formula Weight: 324.41 Flash Point: 110°(230°F) Melting Point: 42-48° UN No.: 3261 MDL No.: MFCD03094651

Formula: C₆H₂BrCl₃O₂S
MW: 324.41 g/mol
Melting Pt: 42…48 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD03094651
CAS Number: 351003-54-8
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR