4-Benzylaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Aminodiphenylmethane
L19873-03
L19873-06
CAAAAL19873-03EA
79.32
CAD
CAAAAL19873-03
CAAAAL19873-06
4-Benzylaniline 98%
4-Benzylaniline
Purity: 98%
CAS No.: 1135-12-2
Molecular Formula: C13H13N
Formula Weight: 183.25
Melting Point: 35-38°
Boiling Point: 142-144°/1mm
MDL No.: MFCD00047861
Formula:
C₁₃H₁₃N MW: 183.25 g/mol Boiling Pt: 142…144 °C/1 mm Melting Pt: 35…38 °C Density: 1.07 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00047861 CAS Number: 1135-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...