2-Chloroisonicotinonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19858-06
L19858-14
CAAAAL19858-06EA
92.11
CAD
CAAAAL19858-06
CAAAAL19858-14
2-Chloroisonicotinonitrile 97%
2-Chloroisonicotinonitrile
Purity: 97%
CAS No.: 33252-30-1
Molecular Formula: C6H3ClN2
Formula Weight: 138.56
Melting Point: 69-72°
Boiling Point: 104-106°/15mm
UN No.: 3439
MDL No.: MFCD00174318
Formula:
C₆H₃ClN₂ Boiling Pt: 104…106 °C/15 mm Melting Pt: 69…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00174318 CAS Number: 33252-30-1 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...