2-Aminoisonicotinonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Aminopyridine-4-carbonitrile, 2-Amino-4-cyanopyridine
L19841-03
L19841-06
L19841-14
CAAAAL19841-03EA
76.16
CAD
CAAAAL19841-03
CAAAAL19841-06
CAAAAL19841-14
2-Aminoisonicotinonitrile 97%
2-Aminoisonicotinonitrile
Purity: 97%
CAS No.: 42182-27-4
Molecular Formula: C6H5N3
Formula Weight: 119.13
Melting Point: 148-152°
Beilstein Reference: 386393
UN No.: 3439
MDL No.: MFCD03791310
Formula:
C₆H₅N₃ MW: 119.13 g/mol Melting Pt: 148…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD03791310 CAS Number: 42182-27-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...