3-Amino-2-bromopyridine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Bromopyridin-3-amine, 2-Bromo-3-pyridylamine
L19839-03
L19839-06
L19839-14
CAAAAL19839-03EA
65.41
CAD
CAAAAL19839-03
CAAAAL19839-06
CAAAAL19839-14
3-Amino-2-bromopyridine 97%
3-Amino-2-bromopyridine
Purity: 98%
CAS No.: 39856-58-1
Molecular Formula: C5H5BrN2
Formula Weight: 173.01
Melting Point: 75-79°
Beilstein Reference: 109830
Formula:
C₅H₅BrN₂ MW: 173.01 g/mol Melting Pt: 75…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234064 CAS Number: 39856-58-1 UN: 2811 ADR: 6.1,III |
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...